(±)-α-Methylbenzylamine ≥99%

Danger

Synonyms: DL-1-Phenylethylamine, (±)-1-Phenylethylamine, (±)-α-Phenylethylamine

M31104-25ML M31104-100ML M31104-500ML
220197-520EA 70.65 USD
220197-520 220197-519 220197-521
(±)-α-Methylbenzylamine ≥99%
(±)-α-Methylbenzylamine
Formula: C₆H₅CH(NH₂)CH₃
MW: 121.18 g/mol
Boiling Pt: 184…186 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008069
CAS Number: 618-36-0
EINECS: 210-545-8
UN: 2735
ADR: 8,II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7749b7c7d0770d82ae5145b1a236458393427163&vid=9ef2ef0e518b58972043eaec1de61438a07bcc37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR