4-Methylbenzophenone

Synonyms: p-Methylbenzophenone, Phenyl(p-tolyl)methanone

M29959-10G M29959-50G
220197-485EA 56.62 USD
220197-485 220197-486
4-Methylbenzophenone
4-Methylbenzophenone
Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 53 - 57 °C
Density: 0.9926 g/cm³ (20 °C)
MDL Number: MFCD00008553
CAS Number: 134-84-9
EINECS: 205-159-1
Merck Index: 12,07470

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR