2-Methylbenzimidazole

Synonyms: 2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole

M29800-100G M29800-5G
220197-480EA 137.85 USD
220197-480 220197-481
2-Methylbenzimidazole
2-Methylbenzimidazole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 175 °C
MDL Number: MFCD00005598
CAS Number: 615-15-6
EINECS: 210-411-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR