L(-)-Menthol ≥98%

Warning

Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol

M2780-10MG M2780-500G-A M2780-100G-A M2780-25G-A
220197-373EA 506.15 USD
220197-373 220197-375 220197-372 220197-374
L(-)-Menthol ≥98%
L(-)-Menthol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS Number: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861

Order Now

Specification Test Results

Assay ~98% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR