4'-Methylacetophenone ≥95%

Warning

Synonyms: Methyl p-tolyl ketone, p-Methylacetophenone, 1-(p-Tolyl)ethanone

M26615-500G M26615-100G
220195-838EA 67.85 USD
220195-838 220195-837
4'-Methylacetophenone ≥95%
4'-Methylacetophenone
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.013 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008751
CAS Number: 122-00-9
EINECS: 204-514-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR