3'-Methylacetophenone ≥98%

Warning

Synonyms: Methyl m-tolyl ketone, 1-(m-Tolyl)ethanone, m-Methylacetophenone, 3′-Methylacetophenone

M26607-25G M26607-5G M26607-100G
220195-835EA 497.54 USD
220195-835 220195-836 220195-834
3'-Methylacetophenone ≥98%
3'-Methylacetophenone
Formula: C₉H₁₀O
MW: 134.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008742
CAS Number: 585-74-0
EINECS: 209-561-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR