2'-Methylacetophenone ≥98%

Synonyms: o-Methylacetophenone, Methyl o-tolyl ketone, 1-(o-Tolyl)ethanone

M26593-5G M26593-25G
220195-832EA 48.14 USD
220195-832 220195-831
2'-Methylacetophenone ≥98%
2'-Methylacetophenone
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 214 °C (1013 hPa)
Density: 1.0262 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008734
CAS Number: 577-16-2
EINECS: 209-408-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR