Melamine

Synonyms: 2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine

M2659-5G M2659-100G M2659-3KG M2659-1KG
220195-830EA 36.92 USD
220195-830 220197-359 220197-361 220197-360
Melamine
Melamine
Formula: C₃H₆N₆
MW: 126.12 g/mol
Density: 1.57 g/cm³ (20 °C)
Flash Pt: >280 °C
MDL Number: MFCD00006055
CAS Number: 108-78-1
EINECS: 203-615-4
Merck Index: 13,05834

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR