1-Amino-3-methoxypropane ≥99%

Synonyms: 3-Methoxypropylamine

M25007-25ML M25007-1L
220196-568EA 24.66 USD
220196-568 220196-567
1-Amino-3-methoxypropane ≥99%
1-Amino-3-methoxypropane
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: < –70 °C
Density: 0.8737 g/cm³ (20 °C)
Flash Pt: 27 °C
MDL Number: MFCD00014831
CAS Number: 5332-73-0
EINECS: 226-241-3
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR