4'-Methoxypropiophenone ≥99%

M24809-25G M24809-5G
220196-564EA 94.77 USD
220196-564 220196-565
4'-Methoxypropiophenone ≥99%
4'-Methoxypropiophenone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: ~ 273 °C (1013 hPa)
Melting Pt: ~ 37.5 °C
Density: ~ 0.94 g/cm³ (20 °C)
Flash Pt: 62 °C
MDL Number: MFCD00009310
CAS Number: 121-97-1
EINECS: 204-512-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR