3-(4-Methoxyphenyl)propionic acid ≥98%

Synonyms: 3-(4-Methoxyphenyl)propanoic acid

M23527-10G M23527-25G
220197-293EA 72.15 USD
220197-293 220197-294
3-(4-Methoxyphenyl)propionic acid ≥98%
3-(4-Methoxyphenyl)propionic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 95 - 102 °C
MDL Number: MFCD00002777
CAS Number: 1929-29-9
EINECS: 217-678-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR