3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

Warning

Synonyms: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

M2128-500MG M2128-100MG M2128-250MG M2128-5G M2128-10G M2128-1G
220197-267EA 129.92 USD
220197-267 220197-263 220197-266 220197-268 220197-264 220197-265
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formula: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1
EINECS: 206-069-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561211eb7a01c80370a0145ebbb045631152528d&vid=201dbd708a0c122ed9fb72129893fc2c816b2632&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR