(±)-Mandelic acid ≥99%

Danger

Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid

M2101-250G M2101-500G
220197-260EA 56.11 USD
220197-260 220197-261
(±)-Mandelic acid ≥99%
(±)-Mandelic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 117…120 °C
Density: 1.361 g/cm³ (20 °C)
MDL Number: MFCD00064250
CAS Number: 90-64-2
EINECS: 210-277-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR