(3-Methoxyphenyl)acetic acid ≥99%

Danger

Synonyms: 3-Methoxyphenylacetic acid

M19007-25G M19007-5G M19007-100G
220197-200EA 136.77 USD
220197-200 220197-201 220197-199
(3-Methoxyphenyl)acetic acid ≥99%
(3-Methoxyphenyl)acetic acid
Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 305.95 °C (1013 hPa)
Melting Pt: 71…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00004334
CAS Number: 1798-09-0
EINECS: 217-282-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR