Disodium malonate monohydrate

M1875-25G M1875-100G M1875-500G
220197-181EA 58.48 USD
220197-181 220197-180 220197-182
Disodium malonate monohydrate
Disodium malonate monohydrate
Formula: C₃H₂Na₂O₄·1H₂O
MW: 166.04 g/mol
MDL Number: MFCD00149114
CAS Number: 26522-85-0
EINECS: 205-514-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR