3-Mercapto-1,2-propanediol ≥97%, Sigma-Aldrich®

Danger

Synonyms: alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol

M1753-500ML M1753-100ML
220197-130EA 920.62 USD
220197-130 220197-129
3-Mercapto-1,2-propanediol ≥97%, Sigma-Aldrich®
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
MW: 108.16 g/mol
Boiling Pt: 118 °C/5 mmHg (lit.)
Density: 1.247 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
Merck Index: 12,09471

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR