5-Methoxy-2-methylindole

Synonyms: 5-Methoxy-2-methyl-1H-indole

M15451-5G M15451-1G
220197-073EA 179.85 USD
220197-073 220197-072
5-Methoxy-2-methylindole
5-Methoxy-2-methylindole
Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 145 °C (1.5 mmHg)
Melting Pt: 89 °C
MDL Number: MFCD00005620
CAS Number: 1076-74-0
EINECS: 214-066-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR