4-Methoxy-o-toluidine ≥98%

Synonyms: 4-Methoxy-2-methylaniline, m-Cresidine, 2-Methyl-p-anisidine, 1-Amino-4-methoxy-2-methylbenzene, 2-Amino-5-methoxytoluene, 4-Amino-3-methylanisole

M15001-25G M15001-5G
220196-082EA 116.31 USD
220196-082 220196-083
4-Methoxy-o-toluidine ≥98%
4-Methoxy-o-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 29…30 °C
Density: 1.065 g/cm³ (20 °C)
MDL Number: MFCD00007735
CAS Number: 102-50-1
EINECS: 203-036-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR