5-Methoxyindole

Warning

Synonyms: 5-Methoxy-1H-indole, Indol-5-yl methyl ether

M14900-1G M14900-5G
220196-076EA 26.92 USD
220196-076 220196-077
5-Methoxyindole
5-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 55 - 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005674
CAS Number: 1006-94-6
EINECS: 213-745-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR