4-Methoxycinnamic acid predominantly trans 99%, Sigma-Aldrich®

Warning

Synonyms: 3-(4-Methoxyphenyl)acrylic acid

M13807-100G M13807-5G M13807-25G
220197-040EA 278.54 USD
220197-040 220197-042 220197-041
4-Methoxycinnamic acid predominantly trans 99%, Sigma-Aldrich®
4-Methoxycinnamic acid
Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: 173.5…174 °C
MDL Number: MFCD00004398
CAS Number: 830-09-1
EINECS: 212-594-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR