4-Methoxybenzenesulfonyl chloride ≥99%

Synonyms: 4-Methoxybenzene-1-sulphonyl chloride

M10204-25G M10204-5G
220196-916EA 88.52 USD
220196-916 220196-917
4-Methoxybenzenesulfonyl chloride ≥99%
4-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 40 - 42 °C
MDL Number: MFCD00007446
CAS Number: 98-68-0
EINECS: 202-692-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR