Sodium molybdate(VI) dihydrate ≥99.5%

M1003-500G M1003-1KG M1003-100G
220196-907EA 121.85 USD
220196-907 220196-906 220196-905
Sodium molybdate(VI) dihydrate ≥99.5%
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
Melting Pt: 687 °C
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR