Ethyl methanesulfonate

M0880-100G M0880-25G M0880-1G M0880-5G M0880-10G
220196-879EA 631.15 USD
220196-879 220196-882 220196-881 220196-883 220196-880
Ethyl methanesulfonate
Ethyl methanesulphonate
Formula: C₃H₈O₃S
MW: 124.16 g/mol
Boiling Pt: 213 °C (1013 hPa)
Density: 1.2079 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007559
CAS Number: 62-50-0
EINECS: 200-536-7
UN: 2810
ADR: 6.1,II
Merck Index: 13,03859

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR