Molybdenum(VI) oxide

Warning

Synonyms: Molybdenum trioxide

M0753-500G M0753-100G
220196-721EA 225.85 USD
220196-721 220196-690
Molybdenum(VI) oxide
Molybdenum(VI) oxide
Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 °C (1013 hPa)
Melting Pt: 795 °C
Density: 4.69 g/cm³ (20 °C)
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 13,06261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR