Methyl cellulose, 1500 mPa.S (2% H₂O)

Synonyms: Cellulose methyl ether

M0387-250G M0387-500G M0387-100G M0387-1KG
220196-536EA 142.77 USD
220196-536 220196-537 220196-534 220196-535
Methyl cellulose, 1500 mPa.S (2% H₂O)
Methyl cellulose
Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR