L(+)-Leucine 98.5-101.0%

Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine

L8912-10MG L8912-25G L8912-1KG L8912-100G
220194-617EA 36.06 USD
220194-617 220194-619 220194-618 220194-616
L(+)-Leucine 98.5-101.0%
L(+)-Leucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 300 °C
Density: 1.165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002617
CAS Number: 61-90-5
EINECS: 200-522-0
Merck Index: 14,05451

Order Now

Specification Test Results

Assay 98.5-101.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR