L(+)-Sodium lactate ≥98%

Synonyms: Sodium (S)-2-hydroxypropanoate, (S)-(+)-2-Hydroxypropanoic acid sodium salt, (S)-(+)-2-Hydroxypropionic acid sodium salt, Sodium-(S)-lactate, (S)-(+)-Lactic acid sodium salt, L(+)-Lactic acid sodium salt

L7022-10G L7022-50G L7022-5G
220195-901EA 155.77 USD
220195-901 220195-902 220195-903
L(+)-Sodium lactate ≥98%
L(+)-Sodium lactate
Formula: C₃H₅NaO₃
MW: 112.06 g/mol
MDL Number: MFCD00066576
CAS Number: 867-56-1
EINECS: 212-762-3

Order Now

Specification Test Results

Assay ~98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR