Lithium acetate dihydrate, Reagent Grade

L6883-1KG L6883-250G
220195-879EA 127.38 USD
220195-879 220195-880
Lithium acetate dihydrate, Reagent Grade
Lithium acetate dihydrate
Formula: H₃CCOOLi·2H₂O
Melting Pt: 291 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR