γ-Linolenic acid methyl ester

Synonyms: Methyl γ-linolenate, Methyl cis,cis,cis-6,9,12-octadecatrienoate, 6Z,9Z,12Z-Octadecatrienoic acid methyl ester, Methyl (6Z,9Z,12Z)-6,9,12-octadecatrienoate, (6Z,9Z,12Z)-Methyl octadeca-6,9,12-trienoate, cis,cis,cis-6,9,12-Octadecatrienoic acid methyl ester

L6503-500MG L6503-1G L6503-100MG
220195-787EA 547.69 USD
220195-787 220195-786 220195-785
γ-Linolenic acid methyl ester
γ-Linolenic acid methyl ester
Formula: C₁₉H₃₂O₂
MW: 292.46 g/mol
MDL Number: MFCD00010455
CAS Number: 16326-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR