Lithium sulfate ≥98.5%

Warning

L6375-100G L6375-500G
220195-769EA 65.08 USD
220195-769 220195-770
Lithium sulfate ≥98.5%
Lithium sulphate
Formula: Li₂O₄S
MW: 109.95 g/mol
Boiling Pt: 1377 °C (1013 hPa)
Melting Pt: 859 °C
Density: 2.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011086
CAS Number: 10377-48-7
EINECS: 233-820-4
Merck Index: 12,05569

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR