DL-Lysine monohydrochloride ≥98%

Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride

L6001-25G L6001-100G L6001-1KG
220195-715EA 69.91 USD
220195-715 220195-713 220195-714
DL-Lysine monohydrochloride ≥98%
DL-Lysine monohydrochloride
Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
Melting Pt: 267 °C
MDL Number: MFCD00064563
CAS Number: 70-53-1
EINECS: 200-739-0

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR