L(+)-Lysine dihydrochloride ≥98%

Synonyms: H-Lys-OH·2HCl, H-Lys-OH.2HCl, (S)-(+)-Lysine dihydrochloride, (S)-(+)-2,6-Diaminohexanoic acid dihydrochloride, (S)-(+)-2,6-Diaminocaproic acid dihydrochloride

L5751-100G L5751-25G
220195-688EA 206.77 USD
220195-688 220195-689
L(+)-Lysine dihydrochloride ≥98%
L(+)-Lysine dihydrochloride
Formula: C₆H₁₆Cl₂N₂O₂
MW: 219.11 g/mol
Melting Pt: 200.2 °C
MDL Number: MFCD00066389
CAS Number: 657-26-1
EINECS: 211-518-3

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR