L-Lysine ≥98%

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

L5501-1G L5501-5G L5501-100G L5501-10MG L5501-25G
220195-301EA 70.28 USD
220195-301 220195-303 220195-299 220195-300 220195-302
L-Lysine ≥98%
L-Lysine
Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 196 °C
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR