Lithium dodecyl sulfate ≥98.5% (by GC)

Warning

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt

L4632-25G L4632-5G L4632-50G
220195-555EA 218.14 USD
220195-555 220195-557 220195-556
Lithium dodecyl sulfate ≥98.5% (by GC)
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2

Order Now

Specification Test Results

Assay 98.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR