Lanthanum(III) oxide

L4000-1KG L4000-100G L4000-500G L4000-250G
220194-408EA 1033.85 USD
220194-408 220194-407 220194-410 220194-409
Lanthanum(III) oxide
Lanthanum(III) oxide
Formula: La₂O₃
MW: 325.81 g/mol
Boiling Pt: 4200 °C (1013 hPa)
Melting Pt: 2315 °C
Density: 6.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR