α-Linolenic acid ethyl ester

Synonyms: Ethyl (9Z,12Z,15Z)-9,12,15-octadecatrienoate, 9Z,12Z,15Z-Octadecatrienoic acid ethyl ester, Ethyl linolenate, (9Z,12Z,15Z)-Ethyl octadeca-9,12,15-trienoate, Linolenic acid ethyl ester, Ethyl α-linolenate, cis,cis,cis-9,12,15-Octadecatrienoic acid ethyl ester, Ethyl cis,cis,cis-9,12,15-octadecatrienoate

L2501-500MG L2501-5G L2501-1G
220195-224EA 224 USD
220195-224 220195-225 220195-223
α-Linolenic acid ethyl ester
α-Linolenic acid ethyl ester
Formula: C₂₀H₃₄O₂
MW: 306.49 g/mol
Boiling Pt: 218 °C (1013 hPa)
Density: 0.8919 g/cm³ (20 °C)
MDL Number: MFCD07371452
CAS Number: 1191-41-9
EINECS: 214-734-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR