γ-Linolenic acid ≥99%

Synonyms: cis,cis,cis-6,9,12-Octadecatrienoic acid, γ-Linolenic acid, (6Z,9Z,12Z)-Octadeca-6,9,12-trienoic acid, 6Z,9Z,12Z-Octadecatrienoic acid

L2378-500MG L2378-10MG L2378-100MG L2378-1G
220195-204EA 612.92 USD
220195-204 220195-202 220195-201 220195-203
γ-Linolenic acid ≥99%
γ-Linolenic acid
Formula: C₁₈H₃₀O₂
MW: 278.44 g/mol
MDL Number: MFCD00065718
CAS Number: 506-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR