D-(-)-Lithium lactate 95-100%

Synonyms: D(-)-Lactic acid lithium salt

L1000-100MG L1000-1G L1000-500MG
220195-036EA 225 USD
220195-036 220195-037 220195-038
D-(-)-Lithium lactate 95-100%
D-(-)-Lithium lactate
Formula: C₃H₅LiO₃
MW: 96.01 g/mol
CAS Number: 27848-81-3
EINECS: 248-693-0

Order Now

Specification Test Results

Assay 95-100%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR