Bole white (kaolin)

K7375-1KG K7375-500G
220194-600EA 68.92 USD
220194-600 220194-601
Bole white (kaolin)
Bole white (kaolin)
Formula: Al₂SiO₇·2H₂O
Melting Pt: 1760 °C
Density: 0.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS Number: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR