2-Oxoglutaric acid

Danger

Synonyms: 2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid

K1750-1KG K1750-100G
220194-513EA 750 USD
220194-513 220194-512
2-Oxoglutaric acid
2-Oxoglutaric acid
Formula: HOOCCH₂CH₂COCOOH
MW: 146.1 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 112 - 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR