L(+)-Isoleucine 98.5-101.0%

Synonyms: L-Isoleucine, (2S,3S)-(+)-Isoleucine, H-Ile-OH

I7403-100G I7403-10MG I7403-25G I7403-1KG
220193-231EA 428.31 USD
220193-231 220193-232 220193-234 220193-233
L(+)-Isoleucine 98.5-101.0%
L(+)-Isoleucine
Formula: H₃CCH₂CH(CH₃)CH(NH₂)COOH
MW: 131.17 g/mol
Melting Pt: 279 - 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00064222
CAS Number: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179

Order Now

Specification Test Results

Assay 98.5-101.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR