3-Iodoaniline ≥98%

Warning

I7209-25G I7209-5G
220193-220EA 92.62 USD
220193-220 220193-221
3-Iodoaniline ≥98%
3-Iodoaniline
Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 24…26 °C
Density: 1.821 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007781
CAS Number: 626-01-7
EINECS: 210-922-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR