2-Iodoaniline ≥98%

Warning

I7004-25G I7004-5G
220193-192EA 127.08 USD
220193-192 220193-193
2-Iodoaniline ≥98%
2-Iodoaniline
Formula: C₆H₆IN
MW: 219.02 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 54 - 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00007680
CAS Number: 615-43-0
EINECS: 210-426-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR