3-Isobutyl-1-methylxanthine

Warning

Synonyms: 2,6-Dihydroxy-3-isobutyl-1-methylpurine, IBMX, 3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione, 3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione

I5879-1G I5879-250MG I5879-5G I5879-100MG
220193-089EA 321.23 USD
220193-089 220193-090 220193-091 220193-088
3-Isobutyl-1-methylxanthine
3-Isobutyl-1-methylxanthine
Formula: C₁₀H₁₄N₄O₂
MW: 222.25 g/mol
Melting Pt: 200…201 °C
MDL Number: MFCD00005584
CAS Number: 28822-58-4
EINECS: 249-259-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR