Indole-5-carboxylic acid

Synonyms: 1H-Indole-5-carboxylic acid

I5400-1G I5400-5G
220189-943EA 53.74 USD
220189-943 220192-561
Indole-5-carboxylic acid
Indole-5-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Freezer
MDL Number: MFCD00005678
CAS Number: 1670-81-1
EINECS: 216-799-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR