myo-Inositol

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

I5125-10MG I5125-1KG I5125-50G I5125-100G I5125-500G
220193-028EA 39.22 USD
220193-028 220193-029 220193-031 220192-466 220193-030
myo-Inositol
myo-Inositol
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 210 °C
Density: 1.752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR