Indole-2-carboxylic acid

Synonyms: 1H-Indole-2-carboxylic acid

I5109-10G I5109-5G I5109-50G
220192-463EA 117.38 USD
220192-463 220192-465 220192-464
Indole-2-carboxylic acid
Indole-2-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 203 - 206 °C
MDL Number: MFCD00005611
CAS Number: 1477-50-5
EINECS: 216-030-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR