Itaconic acid

Danger

Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid

I29204-100G I29204-1KG
220192-718EA 35.32 USD
220192-718 220192-719
Itaconic acid
Itaconic acid
Formula: C₅H₆O₄
MW: 130.1 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 162…167 °C
Density: 1.573 g/cm³ (20 °C)
MDL Number: MFCD00004260
CAS Number: 97-65-4
EINECS: 202-599-6
Merck Index: 13,05264

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR