Isoquinoline ≥97%

I28208-100G I28208-500G I28208-5G
220192-704EA 78.29 USD
220192-704 220192-705 220192-706
Isoquinoline ≥97%
Isoquinoline
Formula: C₉H₇N
MW: 129.16 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 21 - 26 °C
Density: 1.098 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006898
CAS Number: 119-65-3
EINECS: 204-341-8
UN: 2811
ADR: 6.1,II
Merck Index: 13,05242

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR