1-Indanone

Synonyms: 2,3-Dihydro-1H-inden-1-one

I2304-100G I2304-25G I2304-10G
220191-762EA 388.77 USD
220191-762 220191-764 220191-763
1-Indanone
1-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 37…40 °C
Density: 1.1028 g/cm³ (40 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003785
CAS Number: 83-33-0
EINECS: 201-470-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR